siRNA Details
| Gene Symbol: Ryk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042607.1 | NP_001036072.1 | 2 | 342 | ||
| NM_013649.3 | NP_038677.3 | 2 | 342 | |||
| GenBank mRNA | BC032275.1 | - | 2 | 76 | 5370100 | |
| L02210.1 | - | 2 | 317 | |||
| L21707.1 | - | 2 | 352 | |||
| L38394.1 | - | 2 | 265 | |||
| M98547.1 | - | 2 | 306 | |||

Ryk
Genomic Map