siRNA Details
| Gene Symbol: Zkscan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167944.1 | NP_001161416.1 | 1 | 816 | ||
| NM_016683.2 | NP_057892.2 | 1 | 816 | |||
| GenBank mRNA | AB221634.1 | - | 1 | 80 | ||
| AK019484.1 | - | 1 | 807 | |||
| AK173086.1 | - | 1 | 811 | |||
| BC060248.1 | - | 1 | 711 | 5715760 | ||
| U62907.1 | - | 1 | 140 | |||

Zkscan5
Genomic Map