siRNA Details
| Gene Symbol: Cyb561d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019720.4 | NP_062694.1 | 3 | 920 | ||
| GenBank mRNA | AF131206.1 | - | 4 | 792 | ||
| AK030225.1 | - | 3 | 921 | |||
| AK162586.1 | - | 3 | 907 | |||
| BC027212.1 | - | 4 | 789 | 3993231 | ||

Cyb561d2
Genomic Map