siRNA Details
| Gene Symbol: Tsga8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021898.2 | NP_068698.1 | 2 | 967 | ||
| GenBank mRNA | AB032764.1 | - | 2 | 970 | ||
| BC139062.1 | - | 2 | 801 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsga8| Gene Symbol: Tsga8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021898.2 | NP_068698.1 | 2 | 967 | ||
| GenBank mRNA | AB032764.1 | - | 2 | 970 | ||
| BC139062.1 | - | 2 | 801 | |||