siRNA Details
| Gene Symbol: Zfp84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023750.2 | NP_076239.1 | 3 | 286 | ||
| GenBank mRNA | AK014610.1 | - | 3 | 242 | ||
| AK034259.1 | - | 3 | 181 | |||
| AK046223.1 | - | 3 | 233 | |||
| AK083748.1 | - | 3 | 226 | |||
| AK145426.1 | - | 3 | 287 | |||
| AK158422.1 | - | 3 | 236 | |||
| BC008540.1 | - | 3 | 234 | 3495627 | ||

Zfp84
Genomic Map