siRNA Details
| Gene Symbol: Zfp84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023750.2 | NP_076239.1 | 5 | 2042 | ||
| GenBank mRNA | AK014610.1 | - | 5 | 1998 | ||
| AK034259.1 | - | 5 | 1937 | |||
| AK083748.1 | - | 5 | 1983 | |||
| AK145426.1 | - | 6 | 1887 | |||
| AK158422.1 | - | 5 | 1984 | |||

Zfp84
Genomic Map