siRNA Details
| Gene Symbol: Bpifb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025631.3 | NP_079907.2 | 9, 10 | 973 | ||
| GenBank mRNA | AK009346.1 | - | 9, 10 | 945 | ||
| AK009504.1 | - | 9, 10 | 948 | |||
| AK009619.1 | - | 9, 10 | 946 | |||
| AK009754.1 | - | 5, 6 | 549 | |||
| AK009776.1 | - | 9, 10 | 931 | |||
| AK009794.1 | - | 9, 10 | 933 | |||
| AK009871.1 | - | 9, 10 | 942 | |||
| AK010130.1 | - | 9, 10 | 928 | |||
| AK010154.1 | - | 9, 10 | 941 | |||
| AK010157.1 | - | 9, 10 | 972 | |||
| AK010215.1 | - | 9, 10 | 942 | |||
| AK028156.1 | - | 9, 10 | 944 | |||
| AK028158.1 | - | 9, 10 | 947 | |||
| AK075874.1 | - | 9, 10 | 944 | |||
| AK160516.1 | - | 9, 10 | 944 | |||
| BC116681.1 | - | 9, 10 | 928 | 40106489 | ||
| BC116727.1 | - | 9, 10 | 928 | 40106498 | ||

Bpifb2
Genomic Map