siRNA Details
| Gene Symbol: Atp8b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026094.3 | NP_080370.2 | 18 | 2041 | ||
| GenBank mRNA | AY364445.1 | - | 18 | 2041 | ||
| BC118977.1 | - | 18 | 2041 | 40098417 | ||
| BC118978.1 | - | 18 | 2041 | 40098419 | ||

Atp8b3
Genomic Map