siRNA Details
| Gene Symbol: Ssr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026155.3 | NP_080431.1 | 2 | 230 | ||
| GenBank mRNA | AK002818.1 | - | 2 | 231 | ||
| AK029353.1 | - | 2 | 231 | |||
| AK076235.1 | - | 2 | 263 | |||
| AK088555.1 | - | 2 | 235 | |||
| AK150255.1 | - | 2 | 224 | |||
| AK192841.1 | - | 2 | 151 | |||
| AK194871.1 | - | 2 | 151 | |||
| AK199053.1 | - | 2 | 151 | |||
| BC011111.1 | - | 2 | 237 | 3965897 | ||
| BC020335.1 | - | 2 | 202 | |||

Ssr3
Genomic Map