siRNA Details
| Gene Symbol: Ssr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026155.3 | NP_080431.1 | 5 | 881 | ||
| GenBank mRNA | AK002818.1 | - | 5 | 882 | ||
| AK012912.1 | - | 1 | 153 | |||
| AK088555.1 | - | 5 | 886 | |||
| AK150255.1 | - | 5 | 875 | |||
| AK162235.1 | - | 3 | 585 | |||
| BC011111.1 | - | 5 | 888 | 3965897 | ||
| BC020335.1 | - | 5 | 853 | |||

Ssr3
Genomic Map