siRNA Details
| Gene Symbol: U2surp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114977.1 | NP_001108449.1 | 4 | 422 | ||
| NM_026476.2 | NP_080752.2 | 3 | 290 | |||
| GenBank mRNA | AK011588.1 | - | 4 | 310 | ||
| AK011796.1 | - | 4 | 312 | |||
| AK029898.1 | - | 4 | 321 | |||
| BC031497.1 | - | 4 | 285 | |||
| BC068265.1 | - | 3 | 306 | 30022109 | ||
| BC080699.1 | - | 3 | 173 | |||

U2surp
Genomic Map