siRNA Details
| Gene Symbol: U2surp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114977.1 | NP_001108449.1 | 10 | 958 | ||
| NM_026476.2 | NP_080752.2 | 9 | 826 | |||
| GenBank mRNA | AK011796.1 | - | 10 | 845 | ||
| AK029898.1 | - | 10 | 858 | |||
| BC031497.1 | - | 10 | 818 | |||
| BC068265.1 | - | 9 | 842 | 30022109 | ||
| BC080699.1 | - | 9 | 709 | |||

U2surp
Genomic Map