siRNA Details
Gene Symbol: COL4A3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130105.1 | NP_001123577.1 | 3 | 652 | ||
NM_005713.2 | NP_005704.1 | 2 | 553 | |||
NM_031361.2 | NP_112729.1 | 2 | 553 | |||
XM_006714513.2 | XP_006714576.1 | 2 | 553 | |||
XM_011543090.2 | XP_011541392.1 | 2 | 553 | |||
XM_011543091.2 | XP_011541393.1 | 2 | 553 | |||
XM_017008919.1 | XP_016864408.1 | 2 | 553 | |||
GenBank mRNA | AF136450.1 | - | 2 | 539 | ||
AF232930.1 | - | 2 | 521 | |||
AK091851.1 | - | 3 | 443 | |||
AK096854.1 | - | 3 | 809 | |||
AK292087.1 | - | 2 | 553 | |||
AY453385.1 | - | 2 | 131 | |||
AY453386.1 | - | 2 | 131 | |||
BC000102.2 | - | 2 | 537 | 3505746 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
