siRNA Details
| Gene Symbol: Grhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026496.4 | NP_080772.2 | 4 | 666 | ||
| GenBank mRNA | AF411213.1 | - | 4 | 399 | ||
| AK002633.1 | - | 4 | 667 | |||
| BC055035.1 | - | 4 | 455 | 6306106 | ||
| BY703140.1 | - | 4 | 666 | |||

Grhl2
Genomic Map