siRNA Details
| Gene Symbol: Grhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026496.4 | NP_080772.2 | 13, 14 | 1961 | ||
| GenBank mRNA | AF411213.1 | - | 13, 14 | 1694 | ||
| AK161180.1 | - | 6, 7 | 639 | |||
| BC004783.1 | - | 4, 5 | 367 | |||
| BC041795.1 | - | 1, 2 | 94 | |||
| BC042575.1 | - | 8, 9 | 794 | |||
| BC055035.1 | - | 13, 14 | 1750 | 6306106 | ||

Grhl2
Genomic Map