siRNA Details
| Gene Symbol: D10Wsu102e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026579.3 | NP_080855.1 | 4 | 2620 | ||
| GenBank mRNA | AK128995.1 | - | 4 | 2613 | ||
| AK147063.1 | - | 4 | 2603 | |||
| BC035946.1 | - | 4 | 2577 | |||
| BC050904.1 | - | 4 | 2587 | 6416625 | ||
| BC094241.1 | - | 4 | 2582 | 30610591 | ||

D10Wsu102e
Genomic Map