siRNA Details
| Gene Symbol: 9530077C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026739.1 | NP_081015.1 | 2, 3 | 873 | ||
| GenBank mRNA | AK220359.1 | - | 2, 3 | 937 | ||
| BC054761.1 | - | 2, 3 | 873 | 6413638 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530077C05Rik| Gene Symbol: 9530077C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026739.1 | NP_081015.1 | 2, 3 | 873 | ||
| GenBank mRNA | AK220359.1 | - | 2, 3 | 937 | ||
| BC054761.1 | - | 2, 3 | 873 | 6413638 | ||