siRNA Details
| Gene Symbol: 9530077C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026739.1 | NP_081015.1 | 2 | 317 | ||
| GenBank mRNA | AK021129.1 | - | 2 | 353 | ||
| AK160343.1 | - | 2 | 366 | |||
| AK220359.1 | - | 2 | 381 | |||
| BC054761.1 | - | 2 | 317 | 6413638 | ||

9530077C05Rik
Genomic Map