siRNA Details
| Gene Symbol: Rasip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028544.1 | NP_082820.1 | 11 | 2615 | ||
| GenBank mRNA | AK011557.2 | - | 5 | 698 | ||
| AK033276.1 | - | 9 | 1903 | |||
| AK156602.1 | - | 11 | 2627 | |||
| BC072584.1 | - | 8 | 1569 | |||
| BC127059.1 | - | 9 | 1541 | 40109852 | ||
| BC150766.1 | - | 11 | 2615 | 9087677 | ||

Rasip1
Genomic Map