siRNA Details
| Gene Symbol: Polr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027423.1 | NP_081699.1 | 9 | 673 | ||
| GenBank mRNA | AK039244.1 | - | 9 | 708 | ||
| AK039366.1 | - | 9 | 751 | |||
| AK165886.1 | - | 9 | 817 | |||
| BC068143.1 | - | 9 | 673 | 30362434 | ||

Polr3b
Genomic Map