siRNA Details
| Gene Symbol: 4921511H03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027603.2 | NP_081879.1 | 2 | 849 | ||
| GenBank mRNA | AK014870.1 | - | 2 | 1166 | ||
| AK076678.1 | - | 2 | 850 | |||
| AK133311.1 | - | 2 | 882 | |||
| BC049718.1 | - | 2 | 886 | 6742354 | ||

4921511H03Rik
Genomic Map