siRNA Details
| Gene Symbol: Tmem25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027865.2 | NP_082141.1 | 9 | 2207 | ||
| GenBank mRNA | AK002841.1 | - | 9 | 2208 | ||
| AK090375.1 | - | 7 | 2481 | |||
| AK154206.1 | - | 8 | 2693 | |||
| AK165199.1 | - | 9 | 2239 | |||
| AK165827.1 | - | 9 | 2192 | |||
| BC060057.1 | - | 7 | 2530 | 5696102 | ||
| BC060243.1 | - | 7 | 2530 | 5696102 | ||

Tmem25
Genomic Map