siRNA Details
| Gene Symbol: ARPC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005720.3 | NP_005711.1 | 3 | 221 | ||
| XM_006715825.1 | XP_006715888.1 | 3 | 232 | |||
| XM_006715826.1 | XP_006715889.1 | 3 | 216 | |||
| GenBank mRNA | AF006084.1 | - | 3 | 181 | ||
| AK314818.1 | - | 3 | 166 | |||
| AU143525.1 | - | 3 | 221 | |||
| AY007111.1 | - | 1 | 51 | |||
| BC002562.2 | - | 3 | 154 | 3138319 | ||
| BC002988.1 | - | 3 | 190 | 2821335 | ||
| BC007555.1 | - | 3 | 152 | 2967684 | ||

ARPC1B
Genomic Map