siRNA Details
| Gene Symbol: PSKH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006742.2 | NP_006733.1 | 3 | 1633 | ||
| GenBank mRNA | AJ272212.1 | - | 3 | 1592 | ||
| BC021584.1 | - | 1 | 148 | |||
| BC037560.1 | - | 1 | 503 | |||
| BC062616.1 | - | 3 | 1604 | 5188431 | ||

PSKH1
Genomic Map