siRNA Details
| Gene Symbol: TSPAN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005723.3 | NP_005714.2 | 2, 3 | 576 | ||
| GenBank mRNA | AF065389.1 | - | 2, 3 | 488 | ||
| AK295385.1 | - | 2, 3 | 170 | |||
| AK315723.1 | - | 2, 3 | 543 | |||
| BC009704.1 | - | 2, 3 | 474 | 3895933 | ||
| CR541809.1 | - | 2, 3 | 141 | |||
| CR541829.1 | - | 2, 3 | 141 | |||

TSPAN5
Genomic Map