siRNA Details
| Gene Symbol: Rnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028810.2 | NP_083086.1 | 5 | 756 | ||
| GenBank mRNA | AK011442.1 | - | 5 | 754 | ||
| AK035195.1 | - | 5 | 741 | |||
| AK048635.1 | - | 4 | 2291 | |||
| BC009002.1 | - | 5 | 726 | 3500586 | ||

Rnd3
Genomic Map