siRNA Details
| Gene Symbol: Olfr701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028910.3 | NP_083186.2 | 5 | 2020 | ||
| GenBank mRNA | AK016560.1 | - | 5 | 2019 | ||
| BC110461.2 | - | 4 | 1880 | 40039685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr701| Gene Symbol: Olfr701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028910.3 | NP_083186.2 | 5 | 2020 | ||
| GenBank mRNA | AK016560.1 | - | 5 | 2019 | ||
| BC110461.2 | - | 4 | 1880 | 40039685 | ||