siRNA Details
| Gene Symbol: Mrgprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030726.1 | NP_109651.1 | 1 | 1504 | ||
| GenBank mRNA | AF295365.1 | - | 1 | 1504 | ||
| BC116696.1 | - | 1 | 1134 | 40105568 | ||
| BC117525.1 | - | 1 | 1132 | 40105567 | ||
More Information
Set Membership: |
|

Mrgprh
Genomic Map