siRNA Details
| Gene Symbol: Vmn1r56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030740.1 | NP_109665.1 | 1 | 1505 | ||
| GenBank mRNA | AF324868.1 | - | 1 | 1505 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r56| Gene Symbol: Vmn1r56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030740.1 | NP_109665.1 | 1 | 1505 | ||
| GenBank mRNA | AF324868.1 | - | 1 | 1505 | ||