siRNA Details
Gene Symbol: Zfp87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133228.3 | NP_573491.2 | 4 | 1092 | ||
GenBank mRNA | AF242377.1 | - | 4 | 995 | ||
AK028465.1 | - | 1 | 648 | |||
AK030724.1 | - | 4 | 1040 | |||
AK049711.1 | - | 4 | 1012 | |||
BC031988.1 | - | 4 | 995 | 4017149 |
