siRNA Details
| Gene Symbol: Zfp617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133358.3 | NP_579936.2 | 2 | 244 | ||
| GenBank mRNA | AB020542.1 | - | 205 | |||
| AK143880.1 | - | 2 | 218 | |||
| AK146644.1 | - | 2 | 185 | |||
| BC014712.1 | - | 2 | 150 | 4507398 | ||
| BY034417.1 | - | 2 | 233 | |||

Zfp617
Genomic Map