siRNA Details
| Gene Symbol: ARL4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282431.1 | NP_001269360.1 | 1 | 985 | ||
| NM_005737.3 | NP_005728.2 | 1 | 981 | |||
| GenBank mRNA | AB016811.1 | - | 1 | 489 | ||
| AF493892.1 | - | 1 | 518 | |||
| AJ579850.1 | - | 1 | 532 | |||
| AJ579851.1 | - | 1 | 532 | |||
| AK314119.1 | - | 1 | 522 | |||
| BC089043.1 | - | 1 | 985 | 6578402 | ||
| BT019378.1 | - | 1 | 518 | |||
| BT019379.1 | - | 1 | 518 | |||
| Y17804.1 | - | 1 | 532 | |||

ARL4C
Genomic Map