siRNA Details
| Gene Symbol: Csnk1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159591.1 | NP_001153063.1 | 12 | 2141 | ||
| NM_134002.2 | NP_598763.1 | 11 | 2318 | |||
| GenBank mRNA | AK013192.1 | - | 1 | 291 | ||
| AK152413.1 | - | 11 | 2317 | |||
| BC004839.1 | - | 11 | 2260 | 2648652 | ||
| BC005750.1 | - | 9 | 1347 | 3600496 | ||
| BC063083.1 | - | 6 | 4039 | |||
| CF612914.1 | - | 1 | 261 | |||

Csnk1g2
Genomic Map