siRNA Details
| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005740.2 | NP_005731.1 | 4 | 461 | ||
| GenBank mRNA | AK223349.1 | - | 4 | 448 | ||
| AK290566.1 | - | 4 | 414 | |||
| AL035366.1 | - | 4 | 454 | |||
| AY453399.1 | - | 252 | ||||
| BC002968.1 | - | 4 | 428 | 3543962 | ||
| BT009847.1 | - | 3 | 233 | |||
| BX473356.1 | - | 4 | 466 | |||
| CR456487.1 | - | 3 | 318 | |||
| CR542177.1 | - | 3 | 233 | |||
| CR542195.1 | - | 3 | 233 | |||

DNAL4
Genomic Map