siRNA Details
| Gene Symbol: Olfr769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146267.1 | NP_666379.1 | 1 | 921 | ||
| GenBank mRNA | BC119068.1 | - | 1 | 1009 | 8733817 | |
| BC120786.1 | - | 1 | 1009 | 40129709 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr769| Gene Symbol: Olfr769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146267.1 | NP_666379.1 | 1 | 921 | ||
| GenBank mRNA | BC119068.1 | - | 1 | 1009 | 8733817 | |
| BC120786.1 | - | 1 | 1009 | 40129709 | ||