siRNA Details
| Gene Symbol: Glis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147221.2 | NP_671754.2 | 11 | 2818 | ||
| GenBank mRNA | AF220434.1 | - | 9 | 2672 | ||
| AF486579.1 | - | 11 | 2817 | |||
| BC066157.1 | - | 9 | 2253 | 30468539 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glis1| Gene Symbol: Glis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147221.2 | NP_671754.2 | 11 | 2818 | ||
| GenBank mRNA | AF220434.1 | - | 9 | 2672 | ||
| AF486579.1 | - | 11 | 2817 | |||
| BC066157.1 | - | 9 | 2253 | 30468539 | ||