siRNA Details
| Gene Symbol: Unc5cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152823.4 | NP_690036.4 | 2 | 419 | ||
| GenBank mRNA | AK144925.1 | - | 2 | 420 | ||
| AY510108.1 | - | 1 | 286 | |||
| BC095959.1 | - | 2 | 397 | 5349225 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc5cl| Gene Symbol: Unc5cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152823.4 | NP_690036.4 | 2 | 419 | ||
| GenBank mRNA | AK144925.1 | - | 2 | 420 | ||
| AY510108.1 | - | 1 | 286 | |||
| BC095959.1 | - | 2 | 397 | 5349225 | ||