siRNA Details
| Gene Symbol: ALDH6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278594.1 | NP_001265523.1 | 4 | 416 | ||
| NM_005589.3 | NP_005580.1 | 4 | 416 | |||
| XM_017021331.1 | XP_016876820.1 | 2 | 195 | |||
| GenBank mRNA | AF038011.1 | - | 4 | 296 | ||
| AF148505.1 | - | 4 | 359 | |||
| AF159889.1 | - | 4 | 317 | |||
| AJ249994.1 | - | 4 | 347 | |||
| AK222687.1 | - | 4 | 354 | |||
| AK223244.1 | - | 4 | 328 | |||
| AK311478.1 | - | 4 | 372 | |||
| AK312389.1 | - | 4 | 359 | |||
| BC004909.1 | - | 4 | 348 | 3530715 | ||
| BC032371.1 | - | 4 | 416 | 5163797 | ||

ALDH6A1
Genomic Map