siRNA Details
| Gene Symbol: Peli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172835.3 | NP_766423.3 | 7 | 1858 | ||
| GenBank mRNA | AK044418.1 | - | 7 | 1805 | ||
| AK135575.1 | - | 6 | 1706 | |||
| BC028931.1 | - | 7 | 1823 | 5037308 | ||
| BC057046.1 | - | 1 | 2080 | |||
| BY452323.1 | - | 1 | 344 | |||

Peli3
Genomic Map