siRNA Details
| Gene Symbol: BC048403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173022.2 | NP_766610.2 | 2 | 322 | ||
| GenBank mRNA | AK087110.1 | - | 2 | 310 | ||
| AK169891.1 | - | 2 | 312 | |||
| BC065058.1 | - | 2 | 322 | 5720152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC048403| Gene Symbol: BC048403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173022.2 | NP_766610.2 | 2 | 322 | ||
| GenBank mRNA | AK087110.1 | - | 2 | 310 | ||
| AK169891.1 | - | 2 | 312 | |||
| BC065058.1 | - | 2 | 322 | 5720152 | ||