siRNA Details
| Gene Symbol: BC048403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173022.2 | NP_766610.2 | 3 | 1586 | ||
| GenBank mRNA | AK087110.1 | - | 3 | 1575 | ||
| AK166238.1 | - | 3 | 1419 | |||
| AK169891.1 | - | 3 | 1578 | |||
| BC065058.1 | - | 3 | 1586 | 5720152 | ||

BC048403
Genomic Map