siRNA Details
| Gene Symbol: BC048403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173022.2 | NP_766610.2 | 3 | 2786 | ||
| GenBank mRNA | AK166238.1 | - | 5 | 2585 | ||
| AK169891.1 | - | 3 | 2783 | |||
| BC028785.1 | - | 1 | 630 | |||
| BC048403.1 | - | 1 | 708 | |||
| BC065058.1 | - | 3 | 2786 | 5720152 | ||

BC048403
Genomic Map