siRNA Details
| Gene Symbol: Ubtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173784.3 | NP_776145.2 | 3 | 1443 | ||
| GenBank mRNA | AK079381.1 | - | 1 | 737 | ||
| AK165107.1 | - | 782 | ||||
| BC043664.1 | - | 1 | 936 | |||
| BC057026.1 | - | 3 | 1443 | 6825513 | ||

Ubtd2
Genomic Map