siRNA Details
Gene Symbol: 6330549D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032024.1 | - | 2 | 2032 | ||
AK088233.1 | - | 3 | 2432 | |||
BC147330.1 | - | 1 | 855 | |||
BC147331.1 | - | 1 | 855 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6330549D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032024.1 | - | 2 | 2032 | ||
AK088233.1 | - | 3 | 2432 | |||
BC147330.1 | - | 1 | 855 | |||
BC147331.1 | - | 1 | 855 |