siRNA Details
| Gene Symbol: PPP3CC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243974.1 | NP_001230903.1 | 2 | 592 | ||
| NM_001243975.1 | NP_001230904.1 | 2 | 592 | |||
| NM_005605.4 | NP_005596.2 | 2 | 592 | |||
| XM_017013610.1 | XP_016869099.1 | 2 | 385 | |||
| XM_017013611.1 | XP_016869100.1 | 2 | 592 | |||
| GenBank mRNA | AK055013.1 | - | 2 | 743 | ||
| AK299415.1 | - | 2 | 426 | |||
| AY007249.1 | - | 2 | 199 | |||
| BC004864.2 | - | 2 | 479 | 3677098 | ||
| S46622.1 | - | 2 | 438 | |||

PPP3CC
Genomic Map