siRNA Details
Gene Symbol: Gm4763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177593.1 | NP_808261.1 | 3, 4 | 428 | ||
GenBank mRNA | BC037749.1 | - | 3, 4 | 428 | 1429110 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177593.1 | NP_808261.1 | 3, 4 | 428 | ||
GenBank mRNA | BC037749.1 | - | 3, 4 | 428 | 1429110 |