siRNA Details
| Gene Symbol: Serpina7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177920.5 | NP_808588.3 | 5 | 1229 | ||
| GenBank mRNA | AK050361.1 | - | 6 | 774 | ||
| AK149459.1 | - | 5 | 1210 | |||
| AY575783.1 | - | 4 | 1163 | |||
| BC132100.1 | - | 4 | 1224 | 40130377 | ||

Serpina7
Genomic Map