siRNA Details
| Gene Symbol: Mcf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159485.1 | NP_001152957.1 | 6, 7 | 795 | ||
| NM_001159486.1 | NP_001152958.1 | 6, 7 | 795 | |||
| NM_178076.3 | NP_835177.2 | 6, 7 | 698 | |||
| GenBank mRNA | AK048898.1 | - | 6, 7 | 662 | ||
| AK147252.1 | - | 3, 4 | 902 | |||
| AK170186.1 | - | 6, 7 | 756 | |||
| AK170558.1 | - | 6, 7 | 729 | |||
| BC058622.1 | - | 6, 7 | 633 | 5704822 | ||
| DQ978380.1 | - | 5, 6 | 549 | |||
| S76838.1 | - | 7, 8 | 992 | |||

Mcf2l
Genomic Map