siRNA Details
| Gene Symbol: Mcf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159485.1 | NP_001152957.1 | 14, 15 | 1917 | ||
| NM_001159486.1 | NP_001152958.1 | 14, 15 | 1917 | |||
| NM_178076.3 | NP_835177.2 | 14, 15 | 1820 | |||
| GenBank mRNA | AK122259.1 | - | 5, 6 | 725 | ||
| AK147252.1 | - | 11, 12 | 2024 | |||
| AK170186.1 | - | 14, 15 | 1878 | |||
| AK170558.1 | - | 14, 15 | 1851 | |||
| BC058622.1 | - | 14, 15 | 1755 | 5704822 | ||
| DQ978380.1 | - | 13, 14 | 1671 | |||
| S76838.1 | - | 15, 16 | 2114 | |||

Mcf2l
Genomic Map