siRNA Details
| Gene Symbol: Fstl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253719.1 | NP_001240648.1 | 16 | 3021 | ||
| NM_178673.4 | NP_848788.2 | 16 | 3281 | |||
| GenBank mRNA | AF374460.1 | - | 16 | 3255 | ||
| AK033652.1 | - | 16 | 3272 | |||
| AK046070.1 | - | 16 | 3023 | |||
| AK081152.1 | - | 16 | 3272 | |||
| AK122482.1 | - | 7 | 1649 | |||
| AK138388.1 | - | 16 | 3573 | |||
| AK163842.1 | - | 1 | 349 | |||
| BC075640.1 | - | 16 | 3244 | 30545529 | ||

Fstl5
Genomic Map